siRNA Details
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198789.2 | NP_942084.1 | 9 | 1144 | ||
XM_006245000.2 | XP_006245062.2 | 9 | 1143 | |||
GenBank mRNA | BC061580.1 | - | 9 | 1148 | 6890414 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025824, AK200532, AK151919, AK167514, AK029030, and others.
Gene Ontology Categories:
