siRNA Details
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100760.1 | NP_001094230.1 | 5, 6 | 696 | ||
XM_006245244.2 | XP_006245306.1 | 5, 6 | 718 | |||
GenBank mRNA | BC104673.1 | - | 5, 6 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
