siRNA Details
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108807.1 | NP_001102277.1 | 7, 8 | 1036 | ||
XM_006245414.2 | XP_006245476.1 | 6, 7 | 1487 | |||
XM_006245415.2 | XP_006245477.1 | 6, 7 | 1124 |

More Information
Other Species Targeted

Gene Ontology Categories:
