siRNA Details
Gene Symbol: RGD1311447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134587.1 | NP_001128059.1 | 2 | 274 | ||
XM_006245417.2 | XP_006245479.1 | 2 | 3075 | |||
XM_006245418.2 | XP_006245480.1 | 2 | 1759 | |||
XM_008767273.1 | XP_008765495.1 | 2 | 299 | |||
GenBank mRNA | KM657817.1 | - | 2 | 141 |
