siRNA Details
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012204.1 | NP_001012204.1 | 12 | 1564 | ||
XM_006245435.2 | XP_006245497.2 | 11 | 1475 | |||
XM_006245436.2 | XP_006245498.2 | 10 | 1411 | |||
XM_006245437.2 | XP_006245499.2 | 11 | 1486 | |||
GenBank mRNA | BC083645.1 | - | 12 | 1564 | 7135023 |
