siRNA Details
Gene Symbol: Cav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131914.2 | NP_571989.2 | 2 | 370 | ||
GenBank mRNA | AF439780.1 | - | 2 | 247 | ||
BC062059.1 | - | 2 | 370 | 5598269 | ||
FQ209775.1 | - | 2 | 424 |
