siRNA Details
Gene Symbol: Rhox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024901.1 | NP_001020072.1 | 2 | 222 | ||
XM_008773422.1 | XP_008771644.1 | 2 | 324 | |||
GenBank mRNA | DQ058659.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
