siRNA Details
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108813.2 | NP_001102283.2 | 2 | 223 | ||
GenBank mRNA | BC168213.1 | - | 2 | 199 | ||
FM054992.1 | - | 2 | 208 | |||
FQ214979.1 | - | 2 | 184 |
Gene Symbol: NEWGENE_1560955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773489.1 | XP_008771711.1 | 2 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
