siRNA Details
Gene Symbol: LOC100909913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751998.3 | XP_003752046.1 | 2 | 692 |
Gene Symbol: Ndp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108814.1 | NP_001102284.1 | 645 |
