siRNA Details
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108815.1 | NP_001102285.1 | 4 | 547 | ||
GenBank mRNA | BC079470.1 | - | 4 | 525 | ||
BC158792.1 | - | 4 | 566 | 7312115 | ||
EF688600.1 | - | 4 | 525 |

More Information
Other Species Targeted

