siRNA Details
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079470.1 | - | 4, 5 | 589 | ||
BC158792.1 | - | 4, 5 | 630 | 7312115 | ||
EF688600.1 | - | 4, 5 | 589 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079470.1 | - | 4, 5 | 589 | ||
BC158792.1 | - | 4, 5 | 630 | 7312115 | ||
EF688600.1 | - | 4, 5 | 589 |