siRNA Details
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037496.1 | NP_001032585.1 | 6 | 955 | ||
XM_006256737.2 | XP_006256799.1 | 6 | 1605 | |||
XM_006256738.2 | XP_006256800.1 | 6 | 1604 | |||
XM_006256739.2 | XP_006256801.1 | 6 | 1603 | |||
XM_006256740.2 | XP_006256802.1 | 7 | 1475 | |||
XM_006256741.2 | XP_006256803.1 | 7 | 1474 | |||
GenBank mRNA | BC110054.1 | - | 6 | 955 | 7446041 |

More Information
Other Species Targeted

Gene Ontology Categories:
