siRNA Details
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037496.1 | NP_001032585.1 | 5 | 748 | ||
XM_006256737.2 | XP_006256799.1 | 5 | 1398 | |||
XM_006256738.2 | XP_006256800.1 | 5 | 1397 | |||
XM_006256739.2 | XP_006256801.1 | 5 | 1396 | |||
XM_006256740.2 | XP_006256802.1 | 6 | 1268 | |||
XM_006256741.2 | XP_006256803.1 | 6 | 1267 | |||
GenBank mRNA | BC110054.1 | - | 5 | 748 | 7446041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
