siRNA Details
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199502.1 | NP_955796.1 | 7 | 1010 | ||
XM_006257373.2 | XP_006257435.1 | 7 | 1037 | |||
GenBank mRNA | AB080636.1 | - | 7 | 1010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
