siRNA Details
Gene Symbol: RGD1562200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256967.2 | XP_006257029.1 | 4 | 503 | ||
XM_006256968.2 | XP_006257030.1 | 5 | 690 | |||
XM_006256969.2 | XP_006257031.1 | 3 | 335 | |||
XM_008773240.1 | XP_008771462.1 | 4 | 1064 | |||
XM_008773241.1 | XP_008771463.1 | 3 | 311 | |||
XM_343790.6 | XP_343791.1 | 5 | 660 | |||
GenBank mRNA | FQ216301.1 | - | 5 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
