siRNA Details
Gene Symbol: Dmrtc1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014222.2 | NP_001014244.2 | 6, 7 | 805 | ||
GenBank mRNA | BC079417.1 | - | 6, 7 | 791 | 7113562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
