siRNA Details
Gene Symbol: RGD1561958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108819.1 | NP_001102289.1 | 6 | 755 | ||
XM_006257192.2 | XP_006257254.1 | 6 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
