siRNA Details
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191889.1 | NP_001178818.1 | 10 | 1987 | ||
XM_006234247.2 | XP_006234309.1 | 10 | 2282 | |||
XM_006234248.2 | XP_006234310.1 | 10 | 1928 |
