siRNA Details
Gene Symbol: Naa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135839.1 | NP_001129311.1 | 8 | 682 | ||
XM_006229591.2 | XP_006229653.1 | 8 | 992 | |||
XM_006229592.2 | XP_006229654.1 | 8 | 882 | |||
GenBank mRNA | FQ235221.1 | - | 8 | 717 |
