siRNA Details
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127555.1 | NP_001121027.1 | 11, 12 | 1718 | ||
XM_006229598.1 | XP_006229660.1 | 11, 12 | 1701 | |||
GenBank mRNA | BC166780.1 | - | 11, 12 | 1583 | 7934982 |
