siRNA Details
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014223.2 | NP_001014245.2 | 2 | 335 | ||
XM_006229602.1 | XP_006229664.1 | 2 | 390 | |||
GenBank mRNA | BC079074.1 | - | 2 | 291 | 7122158 | |
DQ116785.1 | - | 2 | 335 | |||
FQ232580.1 | - | 2 | 339 |
