siRNA Details
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108825.1 | NP_001102295.1 | 14, 15 | 1630 | ||
XM_008767692.1 | XP_008765914.1 | 14, 15 | 1630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
