siRNA Details
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286183.1 | NP_001273112.1 | 3 | 782 | ||
XM_003750804.2 | XP_003750852.1 | 3 | 910 | |||
XM_003750806.2 | XP_003750854.1 | 2 | 741 | |||
XM_006246153.2 | XP_006246215.1 | 2 | 3080 | |||
XM_006246158.2 | XP_006246220.1 | 3 | 755 | |||
XM_006246159.2 | XP_006246221.1 | 3 | 735 | |||
XM_006246161.1 | XP_006246223.1 | 2 | 669 | |||
XM_008767653.1 | XP_008765875.1 | 3 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
