siRNA Details
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108826.1 | NP_001102296.1 | 6 | 610 | ||
XM_006246303.2 | XP_006246365.1 | 5 | 582 | |||
XM_006246304.2 | XP_006246366.1 | 5 | 558 | |||
XM_006246306.2 | XP_006246368.1 | 5 | 582 | |||
XM_006246310.2 | XP_006246372.1 | 4 | 406 |
