siRNA Details
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108841.1 | NP_001102311.1 | 1021 | |||
XM_008768584.1 | XP_008766806.1 | 5 | 630 | |||
XM_008768585.1 | XP_008766807.1 | 5 | 625 | |||
XM_008768586.1 | XP_008766808.1 | 5 | 625 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008065, AK162675, AK145838, AK139916, AK150455, and others.
Gene Ontology Categories:
