siRNA Details
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025425.1 | NP_001020596.1 | 10 | 1109 | ||
XM_006248768.2 | XP_006248830.1 | 11 | 1294 | |||
XM_006248769.1 | XP_006248831.1 | 11 | 1299 | |||
XM_008768985.1 | XP_008767207.1 | 10 | 1250 | |||
GenBank mRNA | AY879265.1 | - | 10 | 1109 | ||
GU264001.1 | - | 11 | 1157 |
