siRNA Details
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108852.2 | NP_001102322.1 | 1 | 113 | ||
GenBank mRNA | BC162041.1 | - | 1 | 113 | 7100495 | |
FQ226241.1 | - | 1 | 132 | |||
FQ229096.1 | - | 1 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
