siRNA Details
Gene Symbol: Rad9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030042.1 | NP_001025213.1 | 5 | 594 | ||
XM_006249366.2 | XP_006249428.1 | 5 | 1220 | |||
XM_008769241.1 | XP_008767463.1 | 5 | 608 | |||
XM_008769242.1 | XP_008767464.1 | 5 | 612 | |||
XM_008769243.1 | XP_008767465.1 | 4 | 413 | |||
XM_008769244.1 | XP_008767466.1 | 5 | 508 | |||
XM_008769245.1 | XP_008767467.1 | 5 | 447 | |||
XM_008769246.1 | XP_008767468.1 | 5 | 441 | |||
GenBank mRNA | BC099750.1 | - | 5 | 594 | 5599441 |
