siRNA Details
Gene Symbol: Pigc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012207.1 | NP_001012207.1 | 2 | 951 | ||
XM_006250182.2 | XP_006250244.1 | 2 | 1117 | |||
XM_006250183.1 | XP_006250245.1 | 2 | 1017 | |||
XM_008769665.1 | XP_008767887.1 | 2 | 1055 | |||
GenBank mRNA | BC087080.1 | - | 2 | 951 | 7189037 |
