siRNA Details
Gene Symbol: Mael | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108857.1 | NP_001102327.1 | 5, 6 | 606 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175296, AK077103, AK165801, BC050800.
Gene Ontology Categories:
