siRNA Details
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191550.1 | NP_001178479.1 | 2 | 125 | ||
XM_006250249.2 | XP_006250311.1 | 2 | 339 | |||
XM_006250250.2 | XP_006250312.1 | 2 | 337 |
