siRNA Details
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014231.1 | NP_001014253.1 | 10 | 1511 | ||
XM_006250297.2 | XP_006250359.1 | 10 | 1518 | |||
XM_006250298.2 | XP_006250360.1 | 10 | 2138 | |||
XM_006250299.2 | XP_006250361.1 | 10 | 1534 | |||
XM_006250300.2 | XP_006250362.1 | 10 | 1534 | |||
XM_008769746.1 | XP_008767968.1 | 10 | 1534 | |||
GenBank mRNA | BC085957.1 | - | 10 | 1511 | 7134976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
