siRNA Details
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022699.3 | NP_073190.1 | 4 | 342 | ||
GenBank mRNA | BC058471.1 | - | 4 | 346 | 6919868 | |
FQ210232.1 | - | 4 | 339 | |||
FQ210454.1 | - | 4 | 338 | |||
FQ211005.1 | - | 4 | 344 | |||
FQ211389.1 | - | 4 | 340 | |||
FQ211694.1 | - | 4 | 339 | |||
FQ211731.1 | - | 4 | 338 | |||
FQ214466.1 | - | 4 | 339 | |||
FQ217830.1 | - | 4 | 339 | |||
FQ217913.1 | - | 4 | 339 | |||
FQ221046.1 | - | 4 | 339 | |||
FQ221166.1 | - | 4 | 339 | |||
FQ221653.1 | - | 4 | 347 | |||
FQ221662.1 | - | 4 | 340 | |||
FQ221710.1 | - | 4 | 339 | |||
FQ221740.1 | - | 4 | 338 | |||
FQ221836.1 | - | 4 | 339 | |||
FQ222551.1 | - | 4 | 339 | |||
FQ222688.1 | - | 4 | 338 | |||
FQ222703.1 | - | 4 | 338 | |||
FQ223191.1 | - | 4 | 338 | |||
FQ223319.1 | - | 4 | 339 | |||
FQ223321.1 | - | 4 | 320 | |||
FQ223373.1 | - | 4 | 338 | |||
FQ223428.1 | - | 4 | 286 | |||
FQ223604.1 | - | 659 | ||||
FQ223661.1 | - | 4 | 339 | |||
FQ224433.1 | - | 4 | 341 | |||
FQ224506.1 | - | 4 | 338 | |||
FQ224779.1 | - | 4 | 339 | |||
FQ228655.1 | - | 4 | 338 | |||
FQ228991.1 | - | 4 | 338 | |||
FQ229525.1 | - | 4 | 337 | |||
FQ229603.1 | - | 939 | ||||
K02932.1 | - | 3 | 269 |
Gene Symbol: LOC100362027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727709.3 | XP_002727755.1 | 1 | 346 |
Gene Symbol: Rpl30l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_344178.3 | XP_344179.2 | 1 | 269 |

More Information
Other Species Targeted

Gene Ontology Categories:
