siRNA Details
Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037792.1 | NP_001032881.1 | 2 | 178 | ||
XM_006250961.2 | XP_006251023.1 | 2 | 436 | |||
GenBank mRNA | BC100271.1 | - | 2 | 178 | 7445571 | |
FQ229716.1 | - | 2 | 237 | |||
FQ229847.1 | - | 1 | 62 | |||
FQ229969.1 | - | 2 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
