siRNA Details
Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037792.1 | NP_001032881.1 | 2 | 189 | ||
XM_006250961.2 | XP_006251023.1 | 2 | 447 | |||
GenBank mRNA | BC100271.1 | - | 2 | 189 | 7445571 | |
FQ229716.1 | - | 2 | 248 | |||
FQ229847.1 | - | 1 | 73 | |||
FQ229969.1 | - | 2 | 200 |
