siRNA Details
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109278.2 | NP_001102748.1 | 4 | 384 | ||
GenBank mRNA | FN806631.1 | - | 2 | 153 | ||
FN806632.1 | - | 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
