siRNA Details
Gene Symbol: Kcnk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039516.2 | NP_001034605.1 | 5 | 1093 | ||
GenBank mRNA | AM229406.1 | - | 5 | 748 | ||
BC129068.1 | - | 5 | 1093 | 7387039 |

More Information
Other Species Targeted

Gene Ontology Categories:
