siRNA Details
Gene Symbol: Rnase9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008561.1 | NP_001008561.1 | 2 | 488 | ||
XM_006251890.2 | XP_006251952.1 | 2 | 965 | |||
XM_006251891.2 | XP_006251953.1 | 2 | 887 | |||
GenBank mRNA | AY303815.1 | - | 2 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
