siRNA Details
Gene Symbol: Poteg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014238.1 | NP_001014260.1 | 1 | 246 | ||
XM_008771330.1 | XP_008769552.1 | 1 | 295 | |||
GenBank mRNA | BC079270.1 | - | 1 | 246 | 7133714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
