siRNA Details
Gene Symbol: Rnf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253329.2 | XP_006253391.1 | 7, 8 | 957 | ||
XM_006253330.2 | XP_006253392.1 | 7, 8 | 892 | |||
XM_006253331.2 | XP_006253393.1 | 7, 8 | 952 | |||
XM_006253332.2 | XP_006253394.1 | 7, 8 | 961 | |||
XM_344558.7 | XP_344559.5 | 7, 8 | 897 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
