siRNA Details
Gene Symbol: Mrpl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108879.1 | NP_001102349.1 | 2 | 738 | ||
XM_006227762.2 | XP_006227824.1 | 2 | 754 | |||
XM_006227763.2 | XP_006227825.1 | 2 | 748 | |||
XM_006227764.2 | XP_006227826.1 | 2 | 665 | |||
XM_006227765.2 | XP_006227827.1 | 2 | 691 | |||
GenBank mRNA | BC167015.1 | - | 2 | 685 | 9099683 |
Gene Symbol: LOC100910528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748683.3 | XP_003748731.1 | 2 | 754 | ||
XM_006227802.2 | XP_006227864.1 | 2 | 748 | |||
XM_006227803.2 | XP_006227865.1 | 2 | 665 | |||
XM_006227804.2 | XP_006227866.1 | 2 | 691 |

More Information
Other Species Targeted

Gene Ontology Categories:
