siRNA Details
Gene Symbol: Tifab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025029.1 | NP_001020200.1 | 3 | 952 | ||
XM_006253578.2 | XP_006253640.1 | 3 | 1364 | |||
XM_006253579.2 | XP_006253641.1 | 3 | 1253 | |||
XM_008771443.1 | XP_008769665.1 | 2 | 2510 | |||
GenBank mRNA | BC091188.1 | - | 3 | 952 | 7367314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
