siRNA Details
Gene Symbol: Sox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271205.1 | NP_001258134.1 | 1 | 1793 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009238, AK028989, AK053840, BC052736, X70298.
Gene Ontology Categories:
