siRNA Details
Gene Symbol: Psma8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108884.1 | NP_001102354.1 | 2 | 269 |

More Information
Other Species Targeted

Gene Ontology Categories:

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psma8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108884.1 | NP_001102354.1 | 2 | 269 |