siRNA Details
Gene Symbol: Asphd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009716.1 | NP_001009716.1 | 2 | 1263 | ||
XM_008769345.1 | XP_008767567.1 | 3 | 2496 | |||
XM_008769346.1 | XP_008767568.1 | 4 | 2726 | |||
XM_008769347.1 | XP_008767569.1 | 4 | 1686 | |||
XM_008769348.1 | XP_008767570.1 | 4 | 2399 | |||
XM_008769349.1 | XP_008767571.1 | 4 | 2282 | |||
XM_008769350.1 | XP_008767572.1 | 3 | 2184 | |||
XM_008769351.1 | XP_008767573.1 | 2 | 1142 | |||
GenBank mRNA | BC088863.1 | - | 2 | 1263 | 7313227 |

More Information
Other Species Targeted

Gene Ontology Categories:
