siRNA Details
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108895.1 | NP_001102365.1 | 6 | 804 | ||
XM_006255283.2 | XP_006255345.1 | 5 | 938 |

More Information
Other Species Targeted

Gene Ontology Categories:
