siRNA Details
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191557.1 | NP_001178486.1 | 2, 3 | 301 | ||
XM_006255291.2 | XP_006255353.1 | 3, 4 | 1851 |

More Information
Other Species Targeted

