siRNA Details
Gene Symbol: Chtf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194951.1 | NP_001181880.1 | 2 | 148 | ||
GenBank mRNA | AI044989.1 | - | 2 | 148 |

More Information
Other Species Targeted

Gene Ontology Categories:
