siRNA Details
Gene Symbol: Ankrd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772666.1 | XP_008770888.1 | 11 | 7834 | ||
XM_008772667.1 | XP_008770889.1 | 11 | 7831 | |||
XM_008772668.1 | XP_008770890.1 | 11 | 7825 |
