siRNA Details
Gene Symbol: Zfp324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758845.1 | XP_008757067.1 | 2 | 1801 | ||
XM_008758846.1 | XP_008757068.1 | 2 | 1775 | |||
XM_008758847.1 | XP_008757069.1 | 2 | 2128 | |||
XM_344862.7 | XP_344863.6 | 4 | 1517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
